2-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22293.0025 8.22293.0100 8.22293.0025 8.22293.0100
EM8.22293.0025CS 6659.98 USD
CA8.22293.0025 CA8.22293.0100 EM8.22293.0025 EM8.22293.0100
2-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 41 - 43 °C
Molar Mass 151.12 g/mol
Bulk Density 350 kg/m3
Boiling Point 156 °C (20 hPa)
Ignition Point 200 °C (Lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR