1-Methylpyrrolidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Methylpyrrolidine

M-Clarity™ quality level = MQ200

8.16146.0010 8.16146.0100 8.16146.0100 8.16146.0010
EM8.16146.0010CS 37780.4 USD
CA8.16146.0100 CA8.16146.0010 EM8.16146.0010 EM8.16146.0100
1-Methylpyrrolidine for synthesis, Sigma-Aldrich®
1-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
MDL Number: MFCD00003173
CAS Number: 120-94-5
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Specification Test Results

Solubility 213 g/l
Molar Mass 85.15 g/mol
Boiling Point 81 °C
Vapor Pressure 100 hPa (20 °C)
Flash Point -3 °C
Explosion Limit 1.1 - 9.1 %(V)
Density 0.799 g/cm3
pH 11.3 (1 g/l, H2O)
Ignition Point 170 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR