2-Pentanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Methyl Propyl Ketone

M-Clarity™ quality level = MQ200

8.14536.1000 8.14536.2500 8.14536.1000 8.14536.2500
EM8.14536.2500CS 892.88 USD
EM8.14536.2500 CA8.14536.1000 CA8.14536.2500 EM8.14536.1000
2-Pentanone for synthesis, Sigma-Aldrich®
2-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
MDL Number: MFCD00009400
CAS Number: 107-87-9
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Solubility 43 g/l (25 °C)
Melting Point -78 °C
Molar Mass 86.13 g/mol
Boiling Point 101 °C (1013 hPa)
Flash Point 8 °C
Explosion Limit 1.56 - 8.70 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 449 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR