Potassium methanolate 30 - 35% in methanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14222.0500
8.14222.0500
EM8.14222.0500CS
521
USD
CA8.14222.0500
EM8.14222.0500
Potassium methanolate 30 - 35% in methanol for synthesis, Sigma-Aldrich®
Potassium methanolate
Formula:
CH₃KO MW: 70.13 g/mol |
MDL Number:
MFCD00012416 CAS Number: 865-33-8 |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Boiling Point | 95 °C (1013 hPa) |
Vapor Pressure | 36 hPa (20 °C) |
Flash Point | 29.7 °C |
Refractive Index | 1.3654 (20 °C, 589 nm) |
Explosion Limit | 5.5 - 36.5 %(V) Methanol |
Density | 0.99 g/cm3 (20 °C) |
pH | 11 (H2O) |
Ignition Point | 415 °C DIN 51794 |
Ignition Point | 415 °C DIN 51794 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...