2-Pyrrolidone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Pyrrolidin-2-one

M-Clarity™ quality level = MQ200

8.07041.0005 8.07041.0250 8.07041.1000
EM8.07041.0005CS 1493.48 USD
EM8.07041.0005 EM8.07041.0250 EM8.07041.1000
2-Pyrrolidone for synthesis, Sigma-Aldrich®
2-Pyrrolidone
Formula: C₄H₇NO
MDL Number: MFCD00005270
CAS Number: 616-45-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 24 - 26 °C
Molar Mass 85.11 g/mol
Boiling Point 250 °C (1013 hPa)
Vapor Pressure 0.04 hPa (20 °C)
Flash Point 138 °C
Refractive Index 1.48 (20 °C, 589 nm)
Explosion Limit 1.8 - 16.6 %(V)
Density 1.11 g/cm3 (20 °C)
pH 9 - 11 (100 g/l, H2O, 20 °C)
Ignition Point 390 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR