1-Naphthylacetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.06862.0100 8.06862.0500
EM8.06862.0100CS 1235 USD
EM8.06862.0100 EM8.06862.0500
1-Naphthylacetic acid for synthesis, Sigma-Aldrich®
1-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MDL Number: MFCD00004046
CAS Number: 86-87-3

Order Now

Specification Test Results

Solubility 0.42 g/l (20 °C)
Melting Point 125 - 131 °C
Molar Mass 186.21 g/mol
Bulk Density 240 kg/m3
pH 3.0 (0.6 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR