Quinaldine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Methylquinoline

M-Clarity™ quality level = MQ200

8.05805.0100 8.05805.0250
EM8.05805.0100CS 1189 USD
EM8.05805.0100 EM8.05805.0250
Quinaldine for synthesis, Sigma-Aldrich®
Quinaldine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 247.6
Melting Pt: -2
Density: 1.0585 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Specification Test Results

Solubility 2.5 g/l (20 °C)
Melting Point -2 °C
Molar Mass 143.18 g/mol
Boiling Point 246.9 °C
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.6126 (20 °C, 589 nm)
Density 1.06 g/cm3 (20 °C)
pH 6.9 (H2O, 20 °C) (saturated aqueous solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR