Dimethyldichlorosilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03452.0250 8.03452.1000
EM8.03452.0250CS 551 USD
EM8.03452.0250 EM8.03452.1000
Dimethyldichlorosilane for synthesis, Sigma-Aldrich®
Dimethyldichlorosilane
Formula: C₂H₆Cl₂Si
MDL Number: MFCD00000491
CAS Number: 75-78-5
UN: 1162
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point -76 °C
Molar Mass 129.06 g/mol
Boiling Point 70 °C (1013 hPa)
Vapor Pressure  - 200 hPa (20 °C)
Flash Point -5 °C
Refractive Index 1.4038 (20 °C, 589 nm)
Explosion Limit 1.75 - 48.5 %(V)
Density 1.07 g/cm3 (25 °C)
Ignition Point 425 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR