I-Blue Mini Plasmid Kits, IBI Scientific

Supplier: IBI Scientific

IB47170 IB47171 IB47172
89493-624PK 32.65 USD
89493-624 89493-626 89493-628
I-Blue Mini Plasmid Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The I-Blue Mini Plasmid Kit was designed for rapid isolation of plasmid DNA from 1-7ml of cultured bacterial cells. I-Blue Lysis Buffer (an optional color indicator) is included with the kit in order to prevent common handling errors, ensuring efficient cell lysis and neutralization


  • Sample Size: 1 – 7ml bacterial culture
  • Expectant Yield: up to 50µg pure plasmid DNA
  • Operation Time: 20min or less


Specifications
Format: Spin Column
Binding Capacity: 50 µg
Culture Input: 1-7 ml
Culture Type: Cultured bacterial cells
Plasmid Size: 1-15 kb
Typical Yield: up to 50 µg
Elution Volume: 30-100 µl
Operation Time: <15 minutes

Order Now

SPECIFICATIONS

Description I-Blue Mini Plasmid Sample Kit I-Blue Mini Plasmid Kit
Target molecule Plasmid Plasmid
Refine target molecule Plasmid Plasmid
Sample type Bacteria Bacteria
Method / Format Column (silica-based) Column (silica-based)
Prep size Mini Mini
Yield up to 50 µg of pure plasmid DNA up to 50 µg of pure plasmid DNA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR