Powder Funnel with Flat Side, Ace Glass Incorporated
Supplier: Ace Glass
6469-52
89054-510EA 505.67 USD
89054-510
Powder Funnel with Flat Side, Ace Glass Incorporated
Funnels Powder Funnels
Powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks
Fabricated from borosilicate glass.