Copper(II) acetylacetonate ≥98%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

26722-1KG 26722-250G 26722-25G 26722-100G 26722-100G
102584-200EA 474.85 USD
102584-200 102603-948 102626-716 102626-718 CH26722-100G
Copper(II) acetylacetonate ≥98%
Copper (II) acetylacetonate
Formula: C₁₀H₁₄CuO₄
MW: 261.76 g/mol
MDL Number: MFCD00000016
CAS Number: 13395-16-9
New Product

Order Now

Specification Test Results

Molecular Weight 261.76
Synonym Cupric diacetylacetonate 2,4-Pentanedione copper(II)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR