Storage Tube, Micro Schlenk, Ace Glass Incorporated
Supplier: Ace Glass
9703-04 9703-06
89075-446EA 164.91 USD
89075-446 89075-448
Storage Tube, Micro Schlenk, Ace Glass Incorporated
Tubes Test Tubes
This tube is used for storage or simple reactions.
  • Stopcock on side arm is 1 mm bore glass
  • Item supplied with holed cap, O-ring and septa for sealing or syringe retrieval
With a standard taper 10/10 or standard taper 14/10 outer joint externally threaded for making an O-ring-cap-seal connection with mating inner joint.
Order Now
SPECIFICATIONS
Environmentally Preferable