1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine USP, NF
Supplier: AVANTI POLAR LIPIDS INC MS
Synonyms:
L-β-Oleoyl-γ-palmitoyl-α-lecithin
850457C500MG
850457C1G
850457C200MG
850457C25MG
100128-128EA
669.23
USD
100128-128
100126-516
100126-704
100121-930
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine USP, NF
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine
Formula:
C₄₂H₈₂NO₈P MW: 760,091 g/mol |
MDL Number:
MFCD00077387 CAS Number: 26853-31-6 |
Specification Test Results
Molecular Weight | 760.076 |
Exact Mass | 759.578 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...