Sodium peroxodisulfate

Supplier: AQUA SOLUTIONS MS

Synonyms: Disodium peroxodisulphatePeroxydisulphuric acid disodium saltDisodium peroxodisulfateSodium peroxidisulfateSodium peroxidisulphateSodium persulphateSodium persulfatePeroxydisulfuric acid disodium salt

S3400-12KG
101224-462EA 379.93 USD
101224-462
Sodium peroxodisulfate
Sodium peroxodisulphate
Formula: Na₂S₂O₈
MW: 238.11 g/mol
MDL Number: MFCD00003501
CAS Number: 7775-27-1
UN: 1505
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR