Paraformaldehyde, prills, EM Grade, purified
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Polyoxymethylene
Paraformaldehyde is the condensation product of methylene glycol, a substance not so far isolated in its pure form; it is a white solid with a formaldehyde content between 78 and 98% and is supplied either as a Prill or Granular.
All forms contain more than 95.5% paraformaldehyde content. The Prill and Granular forms are small, free-flowing, dust-free beads, the most desired by EM users.
A fast-penetrating EM fixative used in combination with Glutaraldehyde, Acrolein, and Osmium Tetroxide.
Karnovsky, M.J., J. Cell Biol., 27, 137A (1965). Hayat, M.A. Principles & Tech., of EM (Univ. Park Press), 68 (1981).
Formula:
(CH₂O)n Melting Pt: 120 °C (248 °F) Density: 1.46 g/cm³ (12.1837 lbs/gal.) |
MDL Number:
MFCD00133991 CAS Number: 30525-89-4 UN: 2213 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...