N-Ethyldiisopropylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N.N’-Diisopropyl ethylamineN,N-DiisopropylethylamineDIPEAEthyldiisopropylamineHünig’s base

M-Clarity™ quality level = MQ200

8.00894.0250 8.00894.2500 8.00894.0250 8.00894.2500 8.00894.1000 8.00894.1000
EM8.00894.2500CS 3363.79 USD
CA8.00894.1000 CA8.00894.0250 EM8.00894.2500 EM8.00894.1000 EM8.00894.0250 CA8.00894.2500
N-Ethyldiisopropylamine for synthesis, Sigma-Aldrich®
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.76 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Solubility 3.9 g/l (20 °C)
Melting Point -46 °C
Molar Mass 129.24 g/mol
Boiling Point 127 °C (1013 hPa)
Vapor Pressure 16 hPa (20 °C)
Flash Point 9.5 °C
Refractive Index 1.4121 (25 °C, 589 nm)
Explosion Limit 0.7 - 6.3 %(V)
Density 0.76 g/cm3 (20 °C)
pH 12.3 (H2O) (as an emulsion)
Ignition Point 240 °C
Viscosity kinematic 0.88 mm2/s (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR