Copper (II) ethylenediamine 1 mol/L (3 N) in aqueous solution for measuring the viscosity of cellulose acc. to DIN 54270, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: CupriethylendiamineBis(ethylenediamine)copper dihydroxideCopper(II)-ethylenediame complex

1.09288.1000 1.09288.1000
EM1.09288.1000CS 6027.56 USD
EM1.09288.1000 CA1.09288.1000
Copper (II) ethylenediamine 1 mol/L (3 N) in aqueous solution for measuring the viscosity of cellulose acc. to DIN 54270, Supelco®
Copper (II) ethylenediamine
Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.10 g/cm3 (20 °C)
pH 12.0 - 12.5 (10 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR