Trifluoroacetic acid 25% in water for protein sequencing, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: TFA

M-Clarity™ quality level = MQ200

1.08218.0050 1.08218.0050
EM1.08218.0050CS 16729.26 USD
CA1.08218.0050 EM1.08218.0050
Trifluoroacetic acid 25% in water for protein sequencing, Supelco®
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
https://us.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Specification Test Results

Assay (alkalimetric) 24.5 - 25.5 %
Density (d 20\u2007°C/ 4\u2007°C) 1.129 - 1.131
Fluorescence (at 254\u2007nm) ≤ 2 ppb
Fluorescence (at 365\u2007nm) ≤ 2 ppb
Identity (IR-spectrum) passes test
Water (according to Karl Fischer) 74.5 - 75.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR