SP Bel-Art Metal Quick Disconnect Couplings, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19724-0804 F19724-0204
17150-050EA 78.77 USD
17150-050 17150-052 17150-054 17150-056
SP Bel-Art Metal Quick Disconnect Couplings, Bel-Art Products, a part of SP
Fittings and Connectors
Chrome plated brass connectors with Buna-N O-Ring (made by Colder Products) feature easy-to-use slide locks for leak-free, in-line connections, and automatic shut off when disconnected.
  • Easy-to-use design
  • Leakproof in-line connections
  • Automatic shutoff feature when disconnected
  • Barbed ends on hose
Hose barb ends accept flexible tubing.
Operate at temperatures to 82°C (180°F), pressures to 1.7Mpa (250psi), and vacuum to 30 microns of Hg. Chrome plated valves provide dependable operation and are ideal for liquid and gas connectors in biomedical instrumentation and general industrial applications.