High Vacuum Stopcock, Precision Grade, Three-Way Double Oblique Bore, Ace Glass Incorporated
Supplier: Ace Glass
8212-28 8212-26 8212-02 8212-06
89062-740EA 399.35 USD
89062-740 89062-734 89062-736 89062-738
High Vacuum Stopcock, Precision Grade, Three-Way Double Oblique Bore, Ace Glass Incorporated
Stopcocks
Precision grade, with three-way oblique bore hollow or solid glass plug
Plug has hole to permit evacuation of the lower chamber and prevents leakage through the bottom.