High Vacuum Stopcock with Olique Bore, Three-Way, Ace Glass Incorporated
Supplier: Ace Glass
8205-06 8205-05 8205-04
89062-650EA 545.44 USD
89062-650 89062-694 89062-710
High Vacuum Stopcock with Olique Bore, Three-Way, Ace Glass Incorporated
Stopcocks
Three-way oblique bore, with liquid seal
The solid glass plug is bored to permit evacuation of the lower chamber and prevents leakage through the bottom.
Order Now
SPECIFICATIONS
Environmentally Preferable