Sodium amide, crystalline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14095.1000 8.14095.0100 8.14095.1000
EM8.14095.1000CS 1358.57 USD
EM8.14095.1000 CA8.14095.1000 CA8.14095.0100
Sodium amide, crystalline for synthesis, Sigma-Aldrich®
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
MDL Number: MFCD00011117
CAS Number: 7782-92-5
UN: 1390
ADR: 4.3,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 210 °C
Molar Mass 39.01 g/mol
Boiling Point 400 °C (1013 hPa)
Density 1.39 g/cm3 (25 °C)
Ignition Point 450 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR