Dipropylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Propyl-1-propanamine

M-Clarity™ quality level = MQ200

8.03548.1000 8.03548.1000 8.03548.0500 8.03548.0500 8.03548.0100 8.03548.0100
EM8.03548.0500CS 299.73 USD
CA8.03548.0100 CA8.03548.1000 EM8.03548.0500 EM8.03548.1000 EM8.03548.0100 CA8.03548.0500
Dipropylamine for synthesis, Sigma-Aldrich®
Dipropylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 109.4 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.738 g/cm³ (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009362
CAS Number: 142-84-7
EINECS: 205-565-9
UN: 2383
ADR: 3 (8),II
Merck Index: 12,03406

Order Now

Specification Test Results

Solubility 35 g/l (25 °C) (experimental)
Melting Point -63 °C
Molar Mass 101.19 g/mol
Boiling Point 105 °C (1013 hPa)
Vapor Pressure 38 hPa (20 °C)
Flash Point 7 °C
Refractive Index 1.4049 (20 °C, 589 nm)
Explosion Limit 1.8 - 9.3 %(V)
Density 0.74 g/cm3 (20 °C)
pH (1 g/l, H2O, 25 °C)
Ignition Point 260 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR