Vanadium ≥99%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12392.0010 1.12392.0010
EM1.12392.0010CS 16059 USD
EM1.12392.0010 CA1.12392.0010
Vanadium ≥99%, powder, Sigma-Aldrich®
Vanadium
Formula: V
MW: 50.94 g/mol
Density: 6.11 g/cm³ (187 °C)
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1710 °C
Molar Mass 50.94 g/mol
Bulk Density 4000 kg/m3
Boiling Point 3000 °C
Density 6.11 g/cm3 (18.7 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR