Sulfuric acid ≥98.0% (by alkalimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.12080.1000
1.12080.2500
1.12080.2510
1.12080.9025
1.12080.0510
EM1.12080.1000CS
448.35
USD
EM1.12080.1000
EM1.12080.2500
EM1.12080.2510
EM1.12080.9025
EM1.12080.0510
Sulfuric acid ≥98.0% (by alkalimetry), EMSURE® for analysis, Supelco®
Sulfuric acid
For analysis EMSURE®.
Packaging: Safebreak is a robust glass bottle coated with polyethylene (PE). Should the bottle fall and break, all acid and glass splinters are safely retained within the polyethylene coating, thus protecting you from any risk.
Formula:
H₂SO₄ Boiling Pt: ~290 °C (lit.) Melting Pt: –20 °C Density: 1.840 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II Merck Index: 14,08974 |
Specification Test Results
Ag (Silver) | ≤ 0.000002 % |
Al (Aluminium) | ≤ 0.000005 % |
As (Arsenic) | ≤ 0.000001 % |
Assay (alkalimetric) | ≥ 98.0 % |
Ba (Barium) | ≤ 0.000005 % |
Be (Beryllium) | ≤ 0.000001 % |
Bi (Bismuth) | ≤ 0.00001 |
Ca (Calcium) | ≤ 0.00002 % |
Cd (Cadmium) | ≤ 0.000002 % |
Chloride (Cl) | ≤ 0.00001 % |
Co (Cobalt) | ≤ 0.000001 % |
Colour | ≤ 10 Hazen |
Cr (Chromium) | ≤ 0.000005 % |
Cu (Copper) | ≤ 0.000001 % |
Date of expiry | see product label |
Fe (Iron) | ≤ 0.00001 |
Ge (Germanium) | ≤ 0.000005 % |
K (Potassium) | ≤ 0.00001 % |
Li (Lithium) | ≤ 0.000001 % |
Mg (Magnesium) | ≤ 0.000005 % |
Mn (Manganese) | ≤ 0.000001 % |
Mo (Molybdenum) | ≤ 0.000005 % |
Na (Sodium) | ≤ 0.00005 % |
NH₄ (Ammonium) | ≤ 0.0002 % |
Ni (Nickel) | ≤ 0.000002 % |
Nitrate (NO₃) | ≤ 0.00002 % |
Pb (Lead) | ≤ 0.000002 % |
Phosphate (PO₄) | ≤ 0.00005 % |
Residue on ignition | ≤ 0.0005 % |
Se (Selenium) | ≤ 0.0001 % |
Sr (Strontium) | ≤ 0.000002 % |
Substances reducing potassium permanganate (as SO₂) | ≤ 0.0002 % |
Ti (Titanium) | ≤ 0.00001 |
Tl (Thallium) | ≤ 0.000005 % |
V (Vanadium) | ≤ 0.000001 % |
Zn (Zinc) | ≤ 0.000005 % |
Zr (Zirconium) | ≤ 0.00001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...