Glas-Col® Aluminum Housed StirMantle, Ace Glass
Supplier: Ace Glass
12045-12 12045-22 12045-50 12045-55 12045-72 12045-05
89075-974EA 4161.56 USD
89075-974 89075-976 89075-978 89075-980 89075-982 76107-040
Glas-Col® Aluminum Housed StirMantle, Ace Glass
Heaters Heating Mantles
This mantle comes with a built-in magnetic stirrer, able to handle up to 72 L spherical flask and is ideal for continuous distillations
The variable speed control (included) offers total mixing management. DC filtering allows the 1/50 hp motor to run cooler, thus extending motor life. With four bottom, 4 high feet to allow bench clearance and air flow. Stir motor control is detachable and allows speeds of 100–1850 rpm. Temperature range is Ambient 10 to 450°C. Supplied with 0.75" diameter x 3" long magnetic stirring bar. Must be operated through a temperature controller.
Ordering information: 50 L and 72 L Aluminum Housed StirMantles must be hard wired.