Trident Prestained Protein Ladder, GeneTex
Supplier: Genetex
GTX50875
GTX16376
89424-752EA
223.38
USD
89424-752
10793-190
Trident Prestained Protein Ladder, GeneTex
Electrophoresis Reagents
Molecular Weight Markers
The Trident Prestained Protein Ladder is for in vitro laboratory use only.
- Store at 4°C for three months
- Not for any clinical, therapeutic, or diagnostic use in humans or animals
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...