BI-O.K.® Test Pak® Steam Biological Indicators, Propper Manufacturing
Supplier: Propper
26920500 26921000 26922000
58752-801CS 423.19 USD
58752-801 58752-803 58752-805
BI-O.K.® Test Pak® Steam Biological Indicators, Propper Manufacturing
Sterilization Indicators
BI-OK® Test-Pak® features self-contained biological indicators that are designed for routine monitoring and release of loads containing implantable devices in steam sterilization.
  • Self-contained biological indicator vials for steam
  • Vials provide test results in 24 hours
  • Vial design prevents premature activation and minimizes risk of breaking
Vials are inoculated with Bacillus atrophies spores to challenge steam sterilization cycle. After sterilization cycle activate media by crushing the glass vial with the vial activator and incubate at 57 °C. AMore Product Information