Zobell Marine Agar and Broth, HiMedia Laboratories
Supplier: HiMedia
M385-500G
M384-500G
M385-5KG
M385-10KG
M384-2.5KG
M384-5KG
M385-2.5KG
95021-754EA
241.39
USD
95021-754
95021-752
97065-950
97065-952
89225-372
89225-374
89225-376
Zobell Marine Agar and Broth, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
Zobell Marine Agar is used in the cultivation of heterotrophic marine bacteria, while Zobell Marine Broth is used in the isolation and enumeration of heterotrophic marine bacteria.