304 Stainless Steel Beaded Pipe to Male NPT Adapter, Ace Glass Incorporated
Supplier: Ace Glass
8870-02 8870-04 8870-06 8870-08 8870-10 8870-12
89064-788EA 455.06 USD
89064-788 89064-790 89064-792 89064-794 89097-224 89097-226
304 Stainless Steel Beaded Pipe to Male NPT Adapter, Ace Glass Incorporated
Fittings and Connectors
Used to join threaded female fittings, valves and flanges to beaded pipe
Size refers to the bead and the NPT thread. Length refers to the overall length of the adapter.