2-Amino-3-chloro-5-hydroxypyridine
Supplier: CHEM IMPEX INTL INC MS
29220-250MG
29220-5G
29220-1G
102561-356EA
253.43
USD
102561-356
102570-898
102641-438
2-Amino-3-chloro-5-hydroxypyridine
2-Amino-3-chloro-5-hydroxypyridine
Formula:
C₅H₅ClN₂O MW: 144.558 g/mol |
CAS Number:
209328-70-1 |
Specification Test Results
Molecular Formula | C5H5ClN2O |
Molecular Weight | 144.56 |
MDL Number | MFCD09835228 |
Storage Temperature | Store at 0-8°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...