2,5-Dichloroisonicotinic acid ≥95% (by HPLC), Powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

28344-25G 28344-1G 28344-5G
102561-104EA 32.2 USD
102561-104 102610-692 102642-956
2,5-Dichloroisonicotinic acid ≥95% (by HPLC), Powder
2,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00466640
CAS Number: 88912-26-9
EINECS: 000-000-0

Order Now

Specification Test Results

Molecular Formula C6H3Cl2NO2
Molecular Weight 192
MDL Number MFCD00195808
Storage Temperature Store at 0-8°C
Color Off-White to Yellow

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR