2-Amino-3-bromo-5-methylpyridine ≥97%, powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 3-Bromo-5-methylpyridin-2-amine, 2-Amino-3-bromo-5-picoline, 3-Bromo-5-methyl-2-pyridylamine

27806-25G 27806-250G 27806-5G 27806-100G
102570-582EA 68.65 USD
102570-582 102570-584 102585-418 102604-960
2-Amino-3-bromo-5-methylpyridine ≥97%, powder
2-Amino-3-bromo-5-methylpyridine
Formula: C₆H₇BrN₂
MDL Number: MFCD00068231
CAS Number: 17282-00-7

Order Now

Specification Test Results

Molecular Formula C6H7BrN2
Molecular Weight 187.04
MDL Number MFCD00128868
Storage Temperature Store at 0-8°C
Color Yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR