1-Hydroxybenzotriazole hydrate ≥99% (by HPLC) wetted with ca. 20% water, Powder
Supplier: CHEM IMPEX INTL INC MS
24755-100G
24755-250G
24755-10KG
24755-5KG
24755-25G
24755-1KG
24755-25KG
102582-734EA
89.89
USD
102582-734
102582-736
102601-916
102613-720
102625-472
102625-474
102640-784
1-Hydroxybenzotriazole hydrate ≥99% (by HPLC) wetted with ca. 20% water, Powder
1-Hydroxybenzotriazole hydrate
Formula:
C₆H₅N₃O·xH₂O |
MDL Number:
MFCD00005805 CAS Number: 123333-53-9 |
Specification Test Results
Molecular Formula | C6H5N3O·xH2O |
Molecular Weight | 135.12 |
Melting Point | 153-160 ºC |
MDL Number | MFCD00065690 |
Storage Temperature | Store at 0-8°C |
Flash Point | 51 ºC |
Color | White |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...