ß-(4-Hydroxyphenyl)ethyl iodoacetamide 98 HPLC

Supplier: CHEM IMPEX INTL INC MS

Synonyms: beta-(4-Hydroxyphenyl)ethyl iodoacetamide, N- Iodoacetyltyramine

23038-100MG 23038-250MG
102624-528EA 260.89 USD
102624-528 102624-530
ß-(4-Hydroxyphenyl)ethyl iodoacetamide 98 HPLC
ß-(4-Hydroxyphenyl)ethyl iodoacetamide
Formula: C₁₀H₁₂NO₂I
MW: 305.11 g/mol
MDL Number: MFCD01863151
CAS Number: 53527-07-4

Order Now

Specification Test Results

Molecular Formula C10H12NO2I
Molecular Weight 305.12
MDL Number MFCD01863151
Storage Temperature Store at 0-8°C
Color White to Off-White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR