α-Difluoromethyl-DL-ornithine ≥98% (by NMR), Powder
Supplier: CHEM IMPEX INTL INC MS
05771-250MG
05771-1G
102591-934EA
305.91
USD
102591-934
102641-738
α-Difluoromethyl-DL-ornithine ≥98% (by NMR), Powder
α-Difluoromethyl-DL-ornithine
Formula:
C₆H₁₂F₂N₂O₂ |
CAS Number:
70052-12-9 |
Specification Test Results
Molecular Formula | C6H12N2O2F2 |
Molecular Weight | 182.15 |
MDL Number | MFCD00221766 |
Storage Temperature | Store at 0-8°C |
Color | Off-White |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...