N-(2-Chlorotrityl resin)-L-isoleucine benzotriazolyl ester 0,25 - 0,80 meq/g
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
2-Chlorotrityl sesin-L-ile-obt
04825-1G
04825-5G
102562-558EA
277.22
USD
102562-558
102575-920
102591-196
N-(2-Chlorotrityl resin)-L-isoleucine benzotriazolyl ester 0,25 - 0,80 meq/g
N-(2-Chlorotrityl resin)-L-isoleucine benzotriazolyl ester
Specification Test Results
Mesh Size | 200-400 |
Substitution | 0.25-0.80 meq/g |
DVB Percentage | 1% DVB |
MDL Number | MFCD01091425 |
Storage Temperature | Store at 0-8°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...