Na-Boc-Nim-benzyloxymethyl-L-histidine merrifield resin 0,25 - 1,0 meq/g
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
BOC-HIS(BOM)-PAM RESIN
, N-ALPHA-T-BOC-N-IM-BENZYLOXYMETHYL-L-HISTIDINE-PAM RESIN
, BOC-L-HIS(BOM)-MERRIFIELD RESIN
04037-5G
04037-25G
04037-1G
102618-852EA
1196.17
USD
102618-852
102618-850
102618-848
Na-Boc-Nim-benzyloxymethyl-L-histidine merrifield resin 0,25 - 1,0 meq/g
Na-Boc-Nim-benzyloxymethyl-L-histidine merrifield resin
Specification Test Results
Mesh Size | 100-200 |
Substitution | 0.25-1.0 meq/g |
DVB Percentage | 1% DVB |
DVB Percentage | 1% DVB |
MDL Number | MFCD00801284 |
Storage Temperature | Store at 0-8°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...