Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl 0.55 meq/g on Merrifield resin
Supplier: CHEM IMPEX INTL INC MS
02782-5G
02782-25G
02782-1G
102574-792EA
71.14
USD
102574-792
102589-650
102617-802
Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl 0.55 meq/g on Merrifield resin
Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl
Specification Test Results
Mesh Size | 100-200 |
Substitution | 0.55 meq/g |
DVB Percentage | 1% DVB |
MDL Number | MFCD00801209 |
Storage Temperature | Store at 0-8°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...