Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl resin 0,7 meq/g

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Boc-L-Trp(For)-PAM resin

02512-25G 02512-1G 02512-5G 02512-100G
102555-844EA 373.4 USD
102555-844 102617-408 102617-410 102641-570
Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl resin 0,7 meq/g
Na-Boc-Nin-formyl-L-tryptophan 4-oxymethylphenylacetamidomethyl

Order Now

Specification Test Results

Mesh Size 100-200
Substitution 0.7 meq/g
DVB Percentage 1% DVB
MDL Number MFCD00801209
Storage Temperature Store at 0-8°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR