Fmoc-Asn(Trt)-Wang resin 0,514 meq/g
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
(S)-2-(9H-Fluorene-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanal, Fmoc-L-Asn(Trt)-4-methylbenzhydryl resin, Na-Fmoc-Ng-trityl-L-asparagine 4-alkoxybenzyl alcohol resin
01905-1G
01905-5G
01905-25G
102573-960EA
61.03
USD
102573-960
102573-962
102588-792
Fmoc-Asn(Trt)-Wang resin 0,514 meq/g
Fmoc-Asn(Trt)-Wang resin
Specification Test Results
Mesh Size | 100-200 |
Substitution | 0.514 meq/g |
DVB Percentage | 1% DVB |
MDL Number | MFCD00801366 |
Storage Temperature | Store at 0-8°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...