Chloromethyl polystyrene resin, 1.2-1.4 meq/g
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Chloromethylated styrene-divinylbenzene copolymer
, Merrifield Resin
00447-250G
00447-1KG
00447-100G
00447-25G
102572-866EA
418.77
USD
102572-866
102587-606
102587-604
102572-864
Chloromethyl polystyrene resin, 1.2-1.4 meq/g
Chloromethyl polystyrene resin
Chloromethyl polystyrene and styrene monomers are copolymerized in order to obtain the Merrifield resin. This method avoids the use of carcinogenic chloromethyl methyl ether. This resin is used in the synthesis of peptide acids using Boc strategy and can be cleaved using HF or TFMSA. Different Boc protected amino acids attached to this resin are also available from stock. Please inquire.
MDL Number:
MFCD00146406 CAS Number: 55844-94-5 |
Specification Test Results
Mesh Size | 100-200 |
Substitution | 1.2-1.4 meq/g |
DVB Percentage | 1% DVB |
DVB Percentage | 1% DVB |
MDL Number | MFCD00146406 |
Storage Temperature | Store at RT |
Color | Yellow to Pale Yellow |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...