ClickSeal™ Microcentrifuge Tubes, Graduated, National Scientific Supply
Supplier: National Scientific Supply
CN1700-BP CA1700-BP CR1700-BP
20172-698CS 260.94 USD
20172-698 20172-778 20172-900 20172-905 20172-913 20172-920 20172-925 20172-930 20172-946
ClickSeal™ Microcentrifuge Tubes, Graduated, National Scientific Supply
Tubes Microcentrifuge Tubes
Graduated, polypropylene ClickSeal™ Microcentrifuge tubes fit all popular microcentrifuge rotors.
The ClickSeal™ design allows repeated cap closures and eliminates pressure induced popping of caps during incubation. A writing surface is provided on flat cap and side wall for quick sample ID. Tubes withstand temperatures from –80 to 121 °C. This rainbow pack of five bright fluorescent colors includes tubes and caps.