Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport Scientific
T341-2T T341-6T T341-4T T341-5T T341-7T T341-2TPR T341-4TPR T341-5TPR T341-6TPR T341-7TPR T341-2TBR T341-4TBR T341-5TBR T341-6TBR T341-7TBR T341-2TLST T341-4TLST T341-5TLST T341-6TLST T341-7TLST T361-2T T361-4T T361-5T T361-6T T361-7T T361-2TPR T361-4TPR T361-5TPR T361-6TPR T361-7TPR
89050-128CS 140.63 USD
89050-128 60872-412 89050-130 89499-076 89499-078 89499-080 89499-082 89499-084 89499-086 89499-088 89499-090 89499-092 89499-094 89499-096 89499-098 89499-100 89499-102 89499-104 89499-106 89499-108 89499-110 89499-112 89499-114 89499-116 89499-118 89499-120 89499-122 89499-124 89499-126 89499-128 89499-130
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Tubes Microcentrifuge Tubes
Micrewtube™ Microcentrifuge tubes are ideal for freezer storage, boiling application and centrifugation, and will fit most standard microcentrifuge rotors.
  • Made of polypropylene
  • Can be used at temperatures from –196 to 121 °C
Low adhesion Micrewtubes™ Tubes are also available. The specially formulated polypropylene used to manufacture these tubes provides a low adhesion surface to obtain maximum sample yield.
Non-skirted tubes withstand high speed centrifugation up to 20000 ×g. Skirted tubes can withstand up to 17000 ×g. Available plain, graduated,More Product Information