Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Edetic acid disodium saltDisodium dihydrogen ethylenediaminetetraacetateEthylenediaminetetra-acetic acid disodium saltIdranal IIIKestranal® 2SEthylenedinitrilotetra-acetic acid disodium salt

1.09992.0001 1.09992.0001
EM1.09992.0001CS 2337.07 USD
CA1.09992.0001 EM1.09992.0001
Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®
EDTA disodium salt

Concentration after dilution to 1 liter: c(C₁₀H₁₄N₂Na₂O₈ ∙ 2 H₂O) = 0.1 mol/l

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.14 g/cm3 (20 °C)
pH 7.4 - 7.6 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR