Silver nitrate 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Silver mononitrate
, Nitric acid silver salt
, Lunar caustic
M-Clarity™ quality level = MQ100
1.09080.1000
1.09081.4000
1.09081.1000
1.11718.1000
1.09081.9010
1.09081.2500
1.09080.1000
EM1.09081.4000CS
3272
USD
CA1.09080.1000
EM1.09081.4000
EM1.11718.1000
EM1.09081.9010
EM1.09081.2500
EM1.09080.1000
EM1.09081.1000
Silver nitrate 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Silver nitrate
Formula:
AgNO₃ MW: 169.87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1760 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.14 g/cm3 (20 °C) |
pH | 7 - 9 (H2O, 20 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...