Barium sulfate for white standard DIN 5033 (Blanc fixe), Supelco®

Supplier: MilliporeSigma

1.01748.0250 1.01748.0250
EM1.01748.0250CS 3894.43 USD
CA1.01748.0250 EM1.01748.0250
Barium sulfate for white standard DIN 5033 (Blanc fixe), Supelco®
Barium sulphate

Certificate of Guarantee: PTB-Test Official number
Spectral radiance factor is calculated from measurement results
Tristimulus values X,Y,Z and reflection factor R457 are calculated from spectral radiance factor β(λ)d/0

Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1345 °C
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now

Specification Test Results

Spectral radiance factor β(λ)d/₀ (350 nm)
Spectral radiance factor β(λ)d/₀ (360 nm)
Spectral radiance factor β(λ)d/₀ (370 nm)
Spectral radiance factor β(λ)d/₀ (380 nm)
Spectral radiance factor β(λ)d/₀ (390 nm)
Spectral radiance factor β(λ)d/₀ (400 nm)
Spectral radiance factor β(λ)d/₀ (420 nm)
Spectral radiance factor β(λ)d/₀ (450 nm)
Spectral radiance factor β(λ)d/₀ (500 nm)
Spectral radiance factor β(λ)d/₀ (600 nm)
Spectral radiance factor β(λ)d/₀ (700 nm)
Spectral radiance factor β(λ)d/₀ (800 nm)
Spectral radiance factor β(λ)₄₅/₀ (360 nm)
Spectral radiance factor β(λ)₄₅/₀ (370 nm)
Spectral radiance factor β(λ)₄₅/₀ (380 nm)
Spectral radiance factor β(λ)₄₅/₀ (390 nm)
Spectral radiance factor β(λ)₄₅/₀ (400 nm)
Spectral radiance factor β(λ)₄₅/₀ (420 nm)
Spectral radiance factor β(λ)₄₅/₀ (450 nm)
Spectral radiance factor β(λ)₄₅/₀ (500 nm)
Spectral radiance factor β(λ)₄₅/₀ (600 nm)
Spectral radiance factor β(λ)₄₅/₀ (700 nm)
Spectral radiance factor β(λ)₄₅/₀ (800 nm)
Reflection factor R₄₅₇
Standard illuminant A 2° CIE 1931 (X)
Standard illuminant A 2° CIE 1931 (Y)
Standard illuminant A 2° CIE 1931 (Z)
Standard illuminant D65 2° CIE 1931 (X)
Standard illuminant D65 2° CIE 1931 (Y)
Standard illuminant D65 2° CIE 1931 (Z)
Standard illuminant C 2 ° CIE 1931 (X)
Standard illuminant C 2 ° CIE 1931 (Y)
Standard illuminant C 2 ° CIE 1931 (Z)
Standard illuminant A 10° CIE 1964 (X)
Standard illuminant A 10° CIE 1964 (Y)
Standard illuminant A 10° CIE 1964 (Z)
Standard illuminant D65 10 ° CIE 1964 (X)
Standard illuminant D65 10 ° CIE 1964 (Y)
Standard illuminant D65 10 ° CIE 1964 (Z)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR