Sulfuric acid ≥96%, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00714.0250 1.00714.1000
EM1.00714.0250CS 2226 USD
EM1.00714.0250 EM1.00714.1000
Sulfuric acid ≥96%, Suprapur® for trace metal analysis, Supelco®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble,(caution ! development of heat)
Melting Point -20 °C
Molar Mass 98.08 g/mol
Boiling Point 335 °C
Vapor Pressure 0.0001 hPa (20 °C)
Density 1.84 g/cm3 (20 °C)
pH 0.3 (49 g/l, H2O, 25 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR