Double Tube Recrystallizer, Ace Glass Incorporated
Supplier: Ace Glass
7772-11
89060-404EA 535.27 USD
89060-404
Double Tube Recrystallizer, Ace Glass Incorporated
Tubes Recrystallization Tubes
Used for multiple recrystallization of extremely air-sensitive compounds.
  • Approximate volume in each section is 75 ml
A solid crystallized in one arm may be recrystallized in a closed system by decanting the supernatant liquid into the other arm and vacuum distilling the solvent back into the crystals in the first arm. This process may be repeated as often as necessary. After removal of the final supernatant liquid, dried crystals may be transferred to a Schlenk tube. With four standard taper 14/20 outer joints and Porosity B (70 to 100 µm) fritted disc.