Screw Top Headspace Vials, Ace Glass Incorporated
Supplier: Ace Glass
5709-40 5709-30 5709-20 5709-10
89427-760PK 93.64 USD
89427-760 89427-762 89427-758 89427-756
Screw Top Headspace Vials, Ace Glass Incorporated
Closures Chromatography Vial Closures
Headspace style, type 1 borosilicate glass vials with a thread finish top
Accomodates an 18 mm magnetic screw thread cap for autosamplers. Has radius bottom to fit Varian, CTC Analytics and LEAP Technologies Autosamplers. Comes in both clear and amber glass.